Learn More
Abcam Recombinant Dog IL-18 protein (His tag), 100UG
Supplier:  Abcam AB225983100UG
Encompass_Preferred
Recombinant Dog IL-18 protein (His tag) is a Dog Full Length protein, in the 37 to 193 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: IGIF, IL18, Interleukin-18, IL-18, Interferon gamma-inducing factor, Interleukin-1 gamma, IFN-gamma-inducing factor, IL-1 gamma
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2002354