Learn More
Abcam Recombinant EN-TEV Protease protein, 1MG
Supplier:  Abcam AB2859761MG
Encompass_Preferred
Recombinant EN-TEV Protease protein is a Tobacco etch virus Fragment protein, in the 2038 to 2279 aa range, expressed in Escherichia coli, with >=95% purity and suitable for SDS-PAGE, Size Exclusion Chromatography.
Protein/ Peptide/ Enzyme Synonyms: Genome polyprotein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5685