Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Recombinant Endonuclease, Cusativin, 10,000 Units, Lyophilized Bead

Catalog No. NC3863964 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

RapiZyme Cusativin is a recombinant endonuclease designed for advanced digestion and characterization of RNA, including mRNA and sgRNA, prior to LC-MS analysis. It is provided as a lyophilized bead in a skirted tube with LC-MS compatible additives to improve stability and maintain high activity upon reconstitution, and primarily targets 3' Cytidine cleavage for improved sequence coverage.

  • Recombinant endonuclease for advanced RNA digestion and characterization.
  • Supplied as a lyophilized bead in a skirted tube.
  • Contains LC-MS compatible additives for enhanced stability and activity.
  • Primarily targets 3' cytidine cleavage, yielding longer RNA digestion products.
  • Robust and automation-friendly procedure.
  • Supports identity, purity, and critical quality attribute testing of RNA therapeutics.
  • Enables comprehensive sequence mapping and orthogonal oligonucleotide LC-MS maps.

Catalog No. NC3863964 Supplier Waters Corp Supplier No. 186011192
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RapiZyme Cusativin is a recombinant endonuclease designed for advanced digestion and characterization of RNA, including mRNA and sgRNA, prior to LC-MS analysis. It is provided as a lyophilized bead in a skirted tube with LC-MS compatible additives to improve stability and maintain high activity upon reconstitution, and primarily targets 3' Cytidine cleavage for improved sequence coverage.

  • Recombinant endonuclease for advanced RNA digestion and characterization.
  • Supplied as a lyophilized bead in a skirted tube.
  • Contains LC-MS compatible additives for enhanced stability and activity.
  • Primarily targets 3' cytidine cleavage, yielding longer RNA digestion products.
  • Robust and automation-friendly procedure.
  • Supports identity, purity, and critical quality attribute testing of RNA therapeutics.
  • Enables comprehensive sequence mapping and orthogonal oligonucleotide LC-MS maps.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.