Learn More
ILEX LIFE SCIENCES LLC Recombinant Endonuclease Serratia marcescens Expressed in E coli 100000 units 100000 UN 1/EA
SDP
Catalog No. 5000580306
Encompass_Preferred
Ilex Life Sciences Ilexonase Nuclease Recombinant Serratia marcescens Endonuclease 100 000 U DESCRIPTION Ilexonase Nuclease a recombinant endonuclease expressed in E coli is a single non-glycosylated polypeptide chain containing 245 amino acids It has a 30 kDa molecular mass with 2 essential disulfide bonds APPLICATION Ilexonase is a non-specific nuclease that degrades all forms of DNA and RNA (single stranded double stranded linear circular) but has no proteolytic activity This ability has made it a useful enzyme in a variety of biotechnology applications FORMULATION Sterile-filtered colorless liquid solution containing 50% glycerol 50 mM Tris-HCl pH 8 0 20 mM NaCl and 2 mM MgCl2 PURITY 90% (SDS-PAGE) OTHER NAMES broad-spectrum nuclease S marcescens nuclease nuclease A nucA nuc endonuclease EC 3 1 30 2 P13717 Compare to Benzonase Nuclease USAGE Laboratory research use only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80306 Supplier ILEX LIFE SCIENCES LLC Supplier No. A41150100
Ilex Life Sciences Ilexonase Nuclease Recombinant Serratia marcescens Endonuclease 100 000 U DESCRIPTION Ilexonase Nuclease a recombinant endonuclease expressed in E coli is a single non-glycosylated polypeptide chain containing 245 amino acids It has a 30 kDa molecular mass with 2 essential disulfide bonds APPLICATION Ilexonase is a non-specific nuclease that degrades all forms of DNA and RNA (single stranded double stranded linear circular) but has no proteolytic activity This ability has made it a useful enzyme in a variety of biotechnology applications FORMULATION Sterile-filtered colorless liquid solution containing 50% glycerol 50 mM Tris-HCl pH 8 0 20 mM NaCl and 2 mM MgCl2 PURITY 90% (SDS-PAGE) OTHER NAMES broad-spectrum nuclease S marcescens nuclease nuclease A nucA nuc endonuclease EC 3 1 30 2 P13717 Compare to Benzonase Nuclease USAGE Laboratory research use only Not for use in diagnostic or therapeutic procedures