Learn More
Biomatik Corporation Recombinant Escherichia coli Alpha-ketoglutarate-dependent dioxygenase AlkB (alkB)
Supplier:  Biomatik Corporation RPC22689
Encompass_Preferred
Recombinant Escherichia coli Alpha-ketoglutarate-dependent dioxygenase AlkB (alkB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: alkB. Target Synonyms: Alkylated DNA repair protein AlkBDNA oxidative demethylase AlkB. Accession Number: P05050. Expression Region: 1~216aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 44.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5727