Learn More
Biomatik Corporation Recombinant Escherichia coli Lactose operon repressor (lacI)
Supplier:  Biomatik Corporation RPC22816
Encompass_Preferred
Recombinant Escherichia coli Lactose operon repressor (lacI) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: lacI. Target Synonyms: lacI; b0345; JW0336; Lactose operon repressor. Accession Number: P03023. Expression Region: 1~360aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 54.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6227