Learn More
Biomatik Corporation Recombinant Escherichia coli O157:H7 Metalloprotease stcE (stcE), partial
Supplier:  Biomatik Corporation RPC27087
Encompass_Preferred
Recombinant Escherichia coli O157:H7 Metalloprotease stcE (stcE), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli O157:H7. Target Name: stcE. Target Synonyms: MucinaseNeutral zinc metalloprotease StcESecreted protease of C1 esterase inhibitor from EHEC. Accession Number: O82882. Expression Region: 296~551aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 32.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6786