Learn More
R&D Systems™ Recombinant F. meningosepticum PNGase F Protein
Description
Specifications
Specifications
| Accession Number | P21163 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Molecular Weight (g/mol) | M.W. (Observed): 34 kDa, reducing conditions; M.W. (theoretical): 36 kDa |
| Quantity | 100 μg |
| Source | E. coli-derived f. meningosepticum PNGase F protein Ala41-Asn354 with N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Common Name | PNGase F |
| Biological Activity | Measured by its ability to deglycosylate ribonuclease B under denatured conditions. >50% ribonuclease B (10μg) is deglycosylated by 2.5 ng rFmPNGase F within 30 minutes, as measured under the described conditions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.