Learn More
Medchemexpress LLC Recombinant Fc gamma RIIA (CD32a) protein | 50 UG
SDP
Catalog No. 5000292626
Encompass_Preferred
Recombinant Fc gamma RIIA (CD32a) protein derived from rhesus macaque, expressed in HEK293 cells and C-terminally His-tagged. Supplied as 50 μg of purified recombinant protein for use in Fc receptor binding, immune signaling, and protein-protein interaction studies. Suitable for in vitro biochemical and biophysical assays.
Catalog No. 50-002-92626 Supplier Medchemexpress LLC Supplier No. HYP7564750UG
Recombinant Fc gamma RIIA (CD32a) protein derived from rhesus macaque, expressed in HEK293 cells and C-terminally His-tagged. Supplied as 50 μg of purified recombinant protein for use in Fc receptor binding, immune signaling, and protein-protein interaction studies. Suitable for in vitro biochemical and biophysical assays.