Learn More
Abcam Recombinant Firefly Luciferase protein, 500UG
Catalog No. 502585661
Encompass_Preferred
Recombinant Firefly Luciferase protein is a Firefly Full Length protein, in the 1 to 550 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: Luciferin 4-monooxygenase, Luciferase
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5661 Supplier Abcam Supplier No. AB100961500UG
Recombinant Firefly Luciferase protein is a Firefly Full Length protein, in the 1 to 550 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: Luciferin 4-monooxygenase, Luciferase
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement