Learn More
Abcam Recombinant Human TDP43 protein, 20UG
Supplier:  Abcam AB14071820UG
Encompass_Preferred
Recombinant Human TDP43 protein is a Human Fragment protein, in the 1 to 260 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: TDP43, TARDBP, TAR DNA-binding protein 43, TDP-43
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1674404