Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Recombinant HDAC3+NCOR2 complex protein, 50UG

Supplier:  Abcam AB4263150UG

Encompass_Preferred

Recombinant HDAC3+NCOR2 complex protein is a Human protein, expressed in Baculovirus, with >90% purity and suitable for Inhib.

Protein/ Peptide/ Enzyme Synonyms: Histone deacetylase 3, HD3, Protein deacetylase HDAC3, Protein deacylase HDAC3, RPD3-2, SMAP45, HDAC3

Conjugation: Unconjugated

The product is subject to the following: Abcam Restricted Use Statement

Catalog No. 50-267-4123


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.