Learn More
Abcam Recombinant Hepatitis C Virus Core Antigen protein, 100UG
Supplier:  Abcam AB49018100UG
Encompass_Preferred
Recombinant Hepatitis C Virus Core Antigen protein is a Hepatitis C virus (isolate 1) Fragment protein, in the 105 to 302 aa range, expressed in Escherichia coli, with >95% purity and suitable for ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: Genome polyprotein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-4015