Learn More
Active Motif Recombinant Histone H3K4me3 (MLA)
SDP
Supplier:  Active Motif 31600
Encompass
Recombinant human Histone H3 trimethyl Lys4 (H3K4me3) (accession number NP_003520.1) was expressed in E. coli cells and contains two substitutions (cysteine to serine at amino acid 96 and cysteine to alanine at amino acid 110). Recombinant methylated histones are specifically methylated via a chemical alkylation reaction that introduces a methyl lysine analog (MLA). The molecular weight of the protein is 15285 Daltons. Protein Sequence (a.a. substitutions in italics): ARTCQTARKS TGGKAPRKQL ATKAARKSAP ATGGVKKPHR YRPGTVALRE IRRYQKSTEL LIRKLPFQRL VREIAQDFKT DLRFQSSAVM ALQEASEAYL VGLFEDTNLA AIHAKRVTIM PKDIQLARRI RGERA
Catalog No. NC1849018