Learn More
Active Motif Recombinant Histone H3K9me1 (EPL)
SDP
Supplier:  Active Motif 31281
Encompass
Recombinant Histone H3 monomethyl Lys9 (H3K9me1) proteins are generated using expressed protein ligation (EPL) technology. Truncated human Histone H3.2 is produced in E. coli and purified using FPLC. The purified protein is subsequently ligated to a N-terminal histone tail peptide containing monomethyl lysine 9 via a native peptide bond. The full-length protein is then repurified prior to lyophilization. Protein concentration was determined using the molar extinction coefficient for Histone H3 and absorbance at 280nm. The recombinant histone is >98% pure by SDS-PAGE and confirmed by high-resolution ESI-TOF mass spectrometry. The molecular weight of the recombinant histone is 15,239 Daltons. Histone H3 is one of the core components of the nucleosome. The nucleosome is the smallest […] For more information, please visit: https://www.activemotif.com/catalog/513.
Catalog No. NC1844853