Learn More
Abcam Recombinant HIV1 Reverse Transcriptase protein (Active), 200UNIT
Supplier:  Abcam AB63979200UNIT
Encompass_Preferred
Recombinant HIV1 Reverse Transcriptase protein (Active) is a HIV-1 M:B_HXB2R Full Length protein, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, FuncS, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: Gag-Pol polyprotein, Pr160Gag-Pol, gag-pol
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1615552