Learn More
Ibt Bioservices IBT Recombinant Hlg B (tag free), Cat # 1404-001, Staphylococcus aureus, Gamma Hemolysin B, S. aureus, Column chromatography, ELISA, Western Blot, Liquid, Storage: -80°C
SDP
Supplier:  Ibt Bioservices 1404001
Encompass_Preferred
Purified, Staphylococcus aureus recombinant Gamma Hemolysin B (rHlg B). rHlg B (tag free) is expressed in E. coli and purified by FPLC. The theoretical molecular weight of the protein is 35.4 kDa.Supplied in PBS. Store at -20°C for 2-3 weeks, -80°C for long term.
Catalog No. NC2112505