Learn More
Abcam Recombinant Human Nrf2 protein, 10UG
Supplier:  Abcam AB13235610UG
Encompass_Preferred
Recombinant Human Nrf2 protein is a Human Full Length protein, in the 1 to 605 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: NRF2, NFE2L2, Nuclear factor erythroid 2-related factor 2, NF-E2-related factor 2, NFE2-related factor 2, Nrf-2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1733716