Learn More
Abcam Recombinant Human ACE2 protein, 10UG
Supplier:  Abcam AB15185210UG
Encompass_Preferred
Recombinant Human ACE2 protein is a Human Fragment protein, in the 18 to 740 aa range, expressed in HEK 293, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.
Protein/ Peptide/ Enzyme Synonyms: UNQ868/PRO1885, ACE2, Angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, Angiotensin-converting enzyme-related carboxypeptidase, Metalloprotease MPROT15, ACEH, ACE-related carboxypeptidase
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1795129