Learn More
R&D Systems™ Recombinant Human 4-1BB/TNFRSF9 His-tag Protein
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | Q07011 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 3604 |
| Molecular Weight (g/mol) | M.W. (Observed): 25-38 kDa, reducing conditions; M.W. (theoretical): 18 kDa |
| Name | 41BB, 4-1BB, 4-1BB ligand receptor, CD137, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILA, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human 4-1 BB/TNFRSF9/CD137 protein Leu24-His183, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.