Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ACE-2 Fc Chimera Biotinylated Protein

Catalog No. BT10544020
Change view
Click to view available options
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BT10544020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT10544020 Supplier R&D Systems™ Supplier No. BT10544020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA with Recombinant SARS-CoV-2 Spike RBD Fc Chimera (Catalog # 10499-CV ). Measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH (Catalog # ES007 ). The specific activity is >450 pmol/min/μg, as measured under the described conditions.

Specifications

For Use With (Application) Bioactivity, Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, ZnCl2 and Glycerol.
Gene ID (Entrez) 59272
Molecular Weight (g/mol) M.W.-theroretical: 110 kDa, M.W.-observed: 122-134 kDa, under reducing conditions
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human ACE-2 protein Human ACE-2 (Gln18-Ser740) (N-terminus) Accession # Q9BYF1.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Conjugate Biotin
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ACE-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.