Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Activated Coagulation Factor Xa, ACFP

Catalog No. ACFP1063
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACFP1063 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ACFP1063 Supplier R&D Systems™ Supplier No. ACFP1063
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Activated Coagulation Factor Xa, ACFP is derived from Sf 9 (baculovirus). The Recombinant Human Activated Coagulation Factor Xa, ACFP has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P00742
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 2159
Molecular Weight (g/mol) 16.6 kDa & 30 kDa
Quantity 25 μg
Source Spodoptera frugiperda,Sf 9 (baculovirus)-derived human Coagulation Factor X protein Met1-Lys488,with a C-terminal 10-His tag Proform Factor X was expressed,purified,activated and further purified to yield Factor Xa. Produced in an animal component free process (ACFP).
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1 μg of the protein by the LAL method.
Gene Symbol F10
Specific Reactivity Coagulation Factor X
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 25 mM MES, 150 mM NaCl and 5 mM CaCl2, pH 6.0
Buffer Lyophilized from a 0.2 μm filtered solution in MES, NaCl and CaCl2.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.