Learn More
R&D Systems™ Recombinant Human ADAMTS5 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Sodium Acetate, CaCl 2 and NaCl. |
| Gene ID (Entrez) | 11096 |
| Molecular Weight (g/mol) | 41 kDa |
| Name | ADAMTS5 |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line,NS0-derived human ADAMTS5 protein Ser262-Pro622,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -70° C under sterile conditions after reconstitution. |
| Structural Form | Recombinant Human ADAMTS5 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.