Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human adenosine kinase | >95.0% | 50 UG
SDP

Catalog No. 5000292946
Encompass_Preferred

Recombinant human adenosine kinase (ADK) is a full-length, tag-free enzyme produced in HEK293 cells for biochemical and enzymatic research. Supplied as a filtered solution with stabilizers, it offers consistent activity and low endotoxin for reliable assay results.

  • Full-length human sequence (M1-H345).
  • Expressed in HEK293 mammalian cells for native folding.
  • Tag free to minimize assay interference.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Formulated in 20 mM Tris, 300 mM NaCl, pH 8.0, with stabilizers and 20% glycerol.
  • Endotoxin <1 EU/μg and measured specific activity for activity-based assays.
  • Supplied as a frozen solution and shipped on dry ice to preserve activity.

Catalog No. 50-002-92946 Supplier Medchemexpress LLC Supplier No. HYP7553050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human adenosine kinase (ADK) is a full-length, tag-free enzyme produced in HEK293 cells for biochemical and enzymatic research. Supplied as a filtered solution with stabilizers, it offers consistent activity and low endotoxin for reliable assay results.

  • Full-length human sequence (M1-H345).
  • Expressed in HEK293 mammalian cells for native folding.
  • Tag free to minimize assay interference.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Formulated in 20 mM Tris, 300 mM NaCl, pH 8.0, with stabilizers and 20% glycerol.
  • Endotoxin <1 EU/μg and measured specific activity for activity-based assays.
  • Supplied as a frozen solution and shipped on dry ice to preserve activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.