Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Adiponectin/Acrp30 Protein

Catalog No. 1065AP050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1065AP050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1065AP050 Supplier R&D Systems™ Supplier No. 1065AP050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Adiponectin/Acrp30 Protein is derived from NS0. The Recombinant Human Adiponectin/Acrp30 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 9370
Molecular Weight (g/mol) 25.4 kDa
Name Adiponectin/Acrp30
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Adiponectin/Acrp30 protein Glu19-Asn244,with a C-terminal 6-His tag
Storage Requirements Do not freeze. 6 months from date of receipt, 2 to 8° C as supplied.
Structural Form Oligomer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.