Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Aggrecan G1-IGD-G2 Domains Protein

Catalog No. 1220PG025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1220PG025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1220PG025 Supplier R&D Systems™ Supplier No. 1220PG025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity, Binding Activity

The Recombinant Human Aggrecan G1-IGD-G2 Domains Protein is derived from NS0. The Recombinant Human Aggrecan G1-IGD-G2 Domains Protein has been validated for the following applications: Enzyme Activity, Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 176
Molecular Weight (g/mol) 74 kDa
Name Aggrecan G1-IGD-G2 Domains
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived human Aggrecan protein Val20-Gly675,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution For hyaluronan-binding assay, reconstitute at 100 μg/mL in PBS. For use as a metalloprotease substrate, reconstitute at 250 μg/mL in 50 mM Tris, 10 mM CaCl 2 150 mM NaCl and 0.05% Brij-35, pH 7.5.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.