Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Agrin Protein, N-terminal

Catalog No. 8909AG050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8909AG050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8909AG050 Supplier R&D Systems™ Supplier No. 8909AG050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Agrin Protein, N-terminal is derived from CHO. The Recombinant Human Agrin Protein, N-terminal has been validated for the following applications: Bioactivity.

Specifications

Accession Number O00468
For Use With (Application) Bioactivity
Gene ID (Entrez) 375790
Molecular Weight (g/mol) Observed M.W.: 138-181kDa, reducing conditions, Theoretical M.W.: 113kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Agrin protein Thr30-Arg1102,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias agrin, agrin proteoglycan, AGRN
Common Name Agrin
Gene Symbol AGRN
Biological Activity Measured by its ability to inhibit neurite outgrowth of E16-E18 rat embryonic cortical neurons on a Laminin alpha 4 coated plate. Able to significantly inhibit neurite outgrowth when immobilized as a 3μL droplet containing 50 ng of protein.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Thr30-Arg1102, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.