Learn More
R&D Systems™ Recombinant Human Aldehyde Dehydrogenase 3-A1/ALDH3A1
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, Glycerol and DTT. |
| Gene ID (Entrez) | 218 |
| Molecular Weight (g/mol) | 51 kDa |
| Name | Aldehyde Dehydrogenase 3-A1/ALDH3A1 |
| Quantity | 20 μg |
| Source | E. coli-derived human Aldehyde Dehydrogenase 3-A1/ALDH3A1 protein Ser2-His453,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.