Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Angiopoietin-1, Biotinylated Protein

Catalog No. BT923010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT923010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT923010 Supplier R&D Systems™ Supplier No. BT923010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Angiopoietin-1, Biotinylated Protein is derived from NS0. The Recombinant Human Angiopoietin-1, Biotinylated Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q5HYA0
For Use With (Application) Bioactivity
Gene ID (Entrez) 284
Molecular Weight (g/mol) Theoretical M.W.: 56kDa (unlabeled)kDa
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived human Angiopoietin-1 protein Ser20-Phe498,with a C-terminal 6-his tag Accession &Num; Q5HYA0
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, ANGPT1, KIAA0003
Common Name Angiopoietin-1
Gene Symbol ANGPT1
Biological Activity Measured by its ability to inhibit serum deprivation induced apoptosis in HUVEC human umbilical vein endothelial cells. Kwak, H.J. et al. (1999) FEBS Letters 448:249. The ED50 for this effect is typically 10-40ng/mL in the presence of 5μg/mL of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody.
Conjugate Biotin
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Ser20-Phe498, with a C-terminal 6-his tag Accession # Q5HYA0
Purity or Quality Grade >90%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.