Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Angiopoietin-like 4 Biotin Protein

Catalog No. BT8249050
Change view
Click to view available options
:
Quantity: 50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BT8249050 Quantity: 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT8249050 Supplier R&D Systems™ Supplier No. BT8249050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Angiopoietin-like 4 Biotin Protein is derived from CHO. The Recombinant Human Angiopoietin-like 4 Biotin Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number Q9BY76
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CHAPS
Gene ID (Entrez) 51129
Molecular Weight (g/mol) 17 kDa (unlabeled)
Quantity 50μg
Source Chinese Hamster Ovary cell line,CHO-derived human Angiopoietin-like Protein 4/ANGPTL4 protein Gly26-Arg161,with 3xGSL and a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias ANGPTL4, HFARP, HFARPANGPTL2, NL2
Biological Activity 25 to 125ng/mL
Product Type Recombinant protein
Conjugate Biotin
Species Human
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.