Learn More
Biomatik Corporation Recombinant Human Angiotensin-converting enzyme 2 (ACE2), partial, Active Protein
Supplier:  Biomatik Corporation RPC27088
Encompass_Preferred
Recombinant Human Angiotensin-converting enzyme 2 (ACE2), partial, Active Protein is a purified Active Coronavirus Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: <1.0 EU/ug as determined by LAL method. Species: Human (Homo sapiens). Target Name: ACE2. Accession Number: Q9BYF1. Expression Region: 18~740aa. Tag Info: C-Terminal Hfc-Tagged. Theoretical MW: 112.5kDa. Buffer: Lyophilized from a 0.2 um filtered PBS, 6% Trehalose, pH 7.4 Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6181