Learn More
Medchemexpress LLC Recombinant human annexin A5 (ANXA5) | >95.0% | 5 UG
SDP
Catalog No. 5000293580
Encompass_Preferred
Recombinant human Annexin A5 (ANXA5) supplied as a lyophilized protein for research use. This calcium-dependent phospholipid-binding protein is commonly used as an apoptosis marker and in studies of membrane dynamics. Supplied as a 5 μg vial with characterization data for purity and molecular weight.
Catalog No. 50-002-93580 Supplier Medchemexpress LLC Supplier No. HYP7517A5UG
Recombinant human Annexin A5 (ANXA5) supplied as a lyophilized protein for research use. This calcium-dependent phospholipid-binding protein is commonly used as an apoptosis marker and in studies of membrane dynamics. Supplied as a 5 μg vial with characterization data for purity and molecular weight.