Learn More
Abcam Recombinant Human Apolipoprotein E4, 500UG
Supplier:  Abcam AB123766500UG
Encompass_Preferred
Recombinant Human Apolipoprotein E4 is a Human Full Length protein, in the 19 to 317 aa range, expressed in Escherichia coli, with >90% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.
Protein/ Peptide/ Enzyme Synonyms: Apolipoprotein E, Apo-E, APOE
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4844