Learn More
Biomatik Corporation Recombinant Human Apoptosis-associated speck-like protein containing a CARD (PYCARD)
Supplier:  Biomatik Corporation RPC27376
Encompass_Preferred
Recombinant Human Apoptosis-associated speck-like protein containing a CARD (PYCARD) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PYCARD. Target Synonyms: Caspase recruitment domain-containing protein 5PYD and CARD domain-containing proteinTarget of methylation-induced silencing 1. Accession Number: Q9ULZ3. Expression Region: 1~195aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 26.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7923