Learn More
Biomatik Corporation Recombinant Human Apoptosis-associated speck-like protein containing a CARD (PYCARD)
Supplier:  Biomatik Corporation RPC26218
Encompass_Preferred
Recombinant Human Apoptosis-associated speck-like protein containing a CARD (PYCARD) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PYCARD. Target Synonyms: Caspase recruitment domain-containing protein 5 (PYD and CARD domain-containing protein; Target of methylation-induced silencing 1; ASC; CARD5; TMS1). Accession Number: Q9ULZ3. Expression Region: 1~195aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 29.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6622