Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human arylsulfatase A | >95.0% | 100 UG
SDP

Catalog No. 5000290916
Encompass_Preferred

Recombinant human arylsulfatase A is a C-terminal His-tagged enzyme produced in HEK293 cells and supplied lyophilized for enzymatic and biochemical studies. The enzyme hydrolyzes cerebroside sulfate and is commonly assayed using 4-nitrocatechol sulfate (PNCS). Provided with high purity and recommended storage conditions, it is suitable for activity assays and protein characterization.

  • Recombinant human enzyme expressed in HEK293 cells.
  • C-terminal 6xHis tag for purification and detection.
  • Lyophilized formulation from Tris-NaCl buffer for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Biological activity measured with 4-nitrocatechol sulfate (PNCS).
  • Recommend aliquoting and storing at -20 °C or -80 °C for long term.

Catalog No. 50-002-90916 Supplier Medchemexpress LLC Supplier No. HYP7608A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human arylsulfatase A is a C-terminal His-tagged enzyme produced in HEK293 cells and supplied lyophilized for enzymatic and biochemical studies. The enzyme hydrolyzes cerebroside sulfate and is commonly assayed using 4-nitrocatechol sulfate (PNCS). Provided with high purity and recommended storage conditions, it is suitable for activity assays and protein characterization.

  • Recombinant human enzyme expressed in HEK293 cells.
  • C-terminal 6xHis tag for purification and detection.
  • Lyophilized formulation from Tris-NaCl buffer for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Biological activity measured with 4-nitrocatechol sulfate (PNCS).
  • Recommend aliquoting and storing at -20 °C or -80 °C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.