Learn More
R&D Systems™ Recombinant Human Asparagine Synthetase/ASNS His Protein
Description
- Although the name focuses attention on its function in asparagine synthesis, the reaction catalyzed by ASNS also impacts glutamine, aspartate and glutamate homeostasis.
- Although ubiquitously expressed at low levels in many organs, expression is particularly high in the pancreas.
Specifications
Specifications
| Accession Number | P08243-1 |
| For Use With (Application) | Enzyme Activity |
| Gene ID (Entrez) | 440 |
| Molecular Weight (g/mol) | 65 kDa |
| Purification Method | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Asparagine Synthetase/ASNS protein Cys2-Ala561 with C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70°C as supplied. 3 months, -20 to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.