Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human B7-H6 Fc Chimera Biotinylated Protein

Catalog No. BT7144050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT7144050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT7144050 Supplier R&D Systems™ Supplier No. BT7144050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human NKp30 Fc Chimera (Catalog # 1849-NK ) is immobilized at 1 μg/mL, 100 μL/well, the concentration of Biotinylated Recombinant Human B7-H6 Fc Chimera that produces 50% of the optimal binding response is 15-75 ng/mL. Measured by its ability to induce IFN-gamma secretion by NK-92 human natural killer lymphoma cells. Brandt, C.S. et al. (2009) J. Exp. Med. 206:1495. The ED50 for this effect is 0.6-3 μg/mL

Specifications

Accession Number NP_001189368
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 374383
Molecular Weight (g/mol) M.W. (Observed): 74-85 kDa, reducing conditions; M.W. (theoretical): 53 kDa (unlabeled)
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human B7-H6 protein Human B7-H6 (Met1-Ser262) (N-terminus) Accession # NP_001189368 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 2 weeks, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Biotin
Cross Reactivity B7-H6
Species Human
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.