Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human BLOC1S2 protein | ≥95.0% | 100 UG
SDP

Catalog No. 5000294154
Encompass_Preferred

Recombinant human BLOC1S2 protein is supplied as a lyophilized powder expressed in Escherichia coli for research applications. The product corresponds to residues M1-R99, is provided tag free, has an approximate molecular weight of 11.35 kDa and a purity of ≥95.0%. Reconstitute to ≥100 μg/mL in ddH2O and store lyophilized at -20°C; aliquot and freeze for long-term storage.

  • Human protein corresponding to residues M1-R99.
  • Expressed in Escherichia coli.
  • Tag free format.
  • Purity ≥95.0%.
  • Approximate molecular weight 11.35 kDa.
  • Lyophilized powder; reconstitute to ≥100 μg/mL in ddH2O.
  • Stable at -20°C for 2 years; after reconstitution stable 4°C for 1 week or frozen for long term.
  • Intended for biochemical and cell biology research use.

Catalog No. 50-002-94154 Supplier Medchemexpress LLC Supplier No. HYP76171A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human BLOC1S2 protein is supplied as a lyophilized powder expressed in Escherichia coli for research applications. The product corresponds to residues M1-R99, is provided tag free, has an approximate molecular weight of 11.35 kDa and a purity of ≥95.0%. Reconstitute to ≥100 μg/mL in ddH2O and store lyophilized at -20°C; aliquot and freeze for long-term storage.

  • Human protein corresponding to residues M1-R99.
  • Expressed in Escherichia coli.
  • Tag free format.
  • Purity ≥95.0%.
  • Approximate molecular weight 11.35 kDa.
  • Lyophilized powder; reconstitute to ≥100 μg/mL in ddH2O.
  • Stable at -20°C for 2 years; after reconstitution stable 4°C for 1 week or frozen for long term.
  • Intended for biochemical and cell biology research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.