Learn More
Proteintech Group Inc Recombinant Human BMP-2 10ug
SDP
Supplier:  Proteintech Group Inc HZ112810UG
Encompass_Preferred
Animal-free Recombinant Human BMP-2 is expressed from human 293 cells as a disulfide linked homodimeric glycoprotein with an apparent molecular mass of 28 kDa. This cytokine is produced in a serum-free, chemically defined media. Production in human 293 cells offers authentic glycosylation, contributing to stability in cell growth media and other applications. BMPs (Bone Morphogenetic Proteins) belong to the TGF-beta superfamily of structurally related signaling proteins. BMP-2 is a potent osteoinductive cytokine, capable of inducing bone and cartilage formation in association with osteoconductive carriers such as collagen and synthetic hydroxyapatite.
Catalog No. 50-197-3900