Learn More
Proteintech Group Inc Recombinant Human BMP-4 1000ug
SDP
Supplier:  Proteintech Group Inc HZ10451000UG
Encompass_Preferred
Animal-free Recombinant Human BMP-4 is expressed in human 293 cells as a disulfide linked homodimeric glycoprotein with an apparent molecular mass of 34 kDa. This cytokine is produced in a serum-free, chemically defined media.
Catalog No. 50-197-3864
Add to Cart
Request a Quote