Learn More
Proteintech Group Inc Recombinant Human BMP-7 10ug
SDP
Supplier:  Proteintech Group Inc HZ122910UG
Encompass_Preferred
Animal-free Recombinant Human BMP-7 is expressed in human 293 cells as a disulfide linked homodimeric glycoprotein with an apparent molecular mass of 29 kDa. This cytokine is produced in a serum-free, chemically defined media. Recombinant Human BMP-7 is a homodimeric glycoprotein consisting of two 117 amino acid subunits, which correspond to amino acid residues 315 to 431 of the full-length BMP-7 precursor. The protein encoded by this gene is a member of the TGF-beta superfamily. Like other members of the Bone Morphogenetic Protein family of proteins, it plays a key role in the transformation of mesenchymal cells into bone and cartilage.
Catalog No. 50-197-3939