Learn More
Creative Biomart GMP Grade Recombinant Human BMP5 protein
Supplier:  Creative Biomart BMP5221THP
Encompass_Preferred
Recombinant Human BMP5 protein GMP Grade(NP_066551.1)(376 aa) was expressed in HEK293 cells with a C-terminal Fc Tag.
Catalog No. 50-208-5257