Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human BMPR1B/ALK-6 protein (His-GST) | 50 UG
SDP

Catalog No. 5000293907
Encompass_Preferred

Recombinant human BMPR1B (ALK-6) protein (aa 149-502) expressed in Sf9 insect cells with an N-terminal His-GST tag. Intended for research use in biochemical and cell-based assays; provided with low endotoxin and shipped frozen on dry ice.

  • Expressed in Sf9 insect cells to support eukaryotic post-translational modifications.
  • Includes N-terminal His-GST tag for affinity purification and detection.
  • Contains amino acids 149-502 corresponding to the extracellular and kinase domains.
  • Approximate molecular weight 55 kDa for use in analytical and functional assays.
  • Endotoxin level <1 EU/μg to minimize assay interference.
  • Store at -80°C for long-term stability; avoid repeated freeze-thaw cycles.
  • Shipped on dry ice to maintain frozen integrity during transport.

Catalog No. 50-002-93907 Supplier Medchemexpress LLC Supplier No. HYP7617450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human BMPR1B (ALK-6) protein (aa 149-502) expressed in Sf9 insect cells with an N-terminal His-GST tag. Intended for research use in biochemical and cell-based assays; provided with low endotoxin and shipped frozen on dry ice.

  • Expressed in Sf9 insect cells to support eukaryotic post-translational modifications.
  • Includes N-terminal His-GST tag for affinity purification and detection.
  • Contains amino acids 149-502 corresponding to the extracellular and kinase domains.
  • Approximate molecular weight 55 kDa for use in analytical and functional assays.
  • Endotoxin level <1 EU/μg to minimize assay interference.
  • Store at -80°C for long-term stability; avoid repeated freeze-thaw cycles.
  • Shipped on dry ice to maintain frozen integrity during transport.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.