Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human C1qL3 Protein

Catalog No. 9115TN050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9115TN050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9115TN050 Supplier R&D Systems™ Supplier No. 9115TN050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human C1qL3 Protein is derived from E. coli. The Recombinant Human C1qL3 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q5VWW1
For Use With (Application) Bioactivity
Gene ID (Entrez) 389941
Molecular Weight (g/mol) Observed M.W.: 14kDa, reducing conditions, Theoretical M.W.: 16kDa
Quantity 50 μg
Source E. coli-derived human C1qL3 protein Ser122-Asp255,with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias C1ql, C1q-like 3, C1qTNF13, Complement C1q-Like Protein 3, complement component 1, q subcomponent-like 3, CTRP13, K100
Common Name C1QL3
Gene Symbol C1QL3
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human C1qL3 is immobilized at 1μg/mL, 100μL/well, the concentration of Recombinant Human BAI2 Fc Chimera that produces 50% of the optimal binding response is approximately 10-60ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Ser122 - Asp255, with an N-terminal Met and 6-His tag
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.