Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human C1qR1/CD93 (24-580) His Avi Protein, CF

Catalog No. AVI11656050
Change view
Click to view available options
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AVI11656050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AVI11656050 Supplier R&D Systems™ Supplier No. AVI11656050
Only null left
Add to Cart
Add to Cart

His-tag Avi-tag

Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human C1qR1/CD93 (24-580) His-tag Avi-tag binds to Recombinant Human IGFBP-rp1/IGFBP-7 (K95R) Protein (Catalog # 1334-B7 ) with an ED50 of 1.50-15.0 μg/mL.

Specifications

Accession Number Q9NPY3.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 22918
Molecular Weight (g/mol) Observed: 82-105 kDa, under reducing conditions.,Theoretical: 61 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human C1qR1/CD93 protein Ala24-Lys580, with a C-terminal 6-His tag and Avi-tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias AA4 Antigen, C1q R1, C1q receptor 1, C1q Rp, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Collectin Receptor, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Ly68, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Conjugate Biotin
Reconstitution Reconstitute at 200 μg/mL in water.
Protein C1qR1/CD93
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.