Learn More
R&D Systems™ Recombinant Human Cadherin-9 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q9ULB4 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 1007 |
| Molecular Weight (g/mol) | 89.1 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell,HEK293-derived human Cadherin-9 protein, (N-terminus) Human Cadherin-9 (Gly54-Ala615) Accession &Num; Q9ULB4 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Specific Reactivity | Cadherin-9 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.