Learn More
Abcam Recombinant Human Carboxypeptidase B/CPB protein, 10UG
Supplier:  Abcam AB15181210UG
Encompass_Preferred
Recombinant Human Carboxypeptidase B/CPB protein is a Human Full Length protein, in the 16 to 417 aa range, expressed in HEK 293, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.
Protein/ Peptide/ Enzyme Synonyms: CPB, PCPB, CPB1, Carboxypeptidase B, Pancreas-specific protein, PASP
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1561823