Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CART Protein

Catalog No. 10974CO050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10974CO050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10974CO050 Supplier R&D Systems™ Supplier No. 10974CO050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Human CART, immobilized at 5.0 μg/mL on a 96 well plate, is able to significantly enhance neurite outgrowth.

Specifications

Accession Number Q16568.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HCl.
Gene ID (Entrez) 9607
Molecular Weight (g/mol) M.W.-observed: 8-10 kDa, reducing conditions., M.W.-theroretical: 9 kDa
Quantity 50 μg
Source E. coli-derived human CART protein Ala37-Leu116
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CART prepropeptide, CARTcocaine and amphetamine regulated transcript, CARTPT, cocaine- and amphetamine-regulated transcript protein
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CART/CARTPT
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.