Learn More
Abcam Recombinant human Catalase protein, 100UG
Catalog No. 502658225
Encompass_Preferred
Recombinant human Catalase protein is a Human Full Length protein, in the 1 to 527 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Catalase, CAT
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-8225 Supplier Abcam Supplier No. AB130029100UG
Recombinant human Catalase protein is a Human Full Length protein, in the 1 to 527 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Catalase, CAT
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement